RIE156323100 Rieke Metals, Inc.

2-Heptanoylpyridine

  • MDL number MFCD00778377
  • Cas Number : 60975-82-8
  • Molecular Weight : 191.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5569-01E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-01E
Availability details for
Quantity
  • Availability
  • Add to Cart