RIE156323107 Rieke Metals, Inc.

2-(3-Methoxybenzoyl)pyridine

  • MDL number MFCD07699224
  • Cas Number : 55030-49-4
  • Molecular Weight : 213.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5570-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-03
Availability details for
Quantity
  • Availability
  • Add to Cart