RIE156323118 Rieke Metals, Inc.

2-(3,4-Dichlorobenzoyl)pyridine

  • MDL number MFCD07699270
  • Cas Number : 95898-75-2
  • Molecular Weight : 252.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5570-47

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-47
Availability details for
Quantity
  • Availability
  • Add to Cart