RIE156323122 Rieke Metals, Inc.

cyclopentyl phenyl ketone

  • MDL number MFCD00001378
  • Cas Number : 5422-88-8
  • Molecular Weight : 174.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-03-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-03-01
Availability details for
Quantity
  • Availability
  • Add to Cart