RIE156323124 Rieke Metals, Inc.

3-Chloro-4-pyridyl isopropyl ketone

  • MDL number MFCD07699446
  • Cas Number : 898785-53-0
  • Molecular Weight : 183.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-chloro-4-pyridinyl)-2-methyl-1-propanone
Inchi Code 1S/C9H10ClNO/c1-6(2)9(12)7-3-4-11-8(10)5-7/h3-6H,1-2H3
Inchi Key WGVKTECOAYHELX-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5574-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-10
Availability details for
Quantity
  • Availability
  • Add to Cart