RIE156323127 Rieke Metals, Inc.

2-(2,6-Dimethoxybenzoyl)pyridine

  • MDL number MFCD07699274
  • Cas Number : 52856-16-3
  • Molecular Weight : 243.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5570-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-52
Availability details for
Quantity
  • Availability
  • Add to Cart