RIE156323128 Rieke Metals, Inc.

Cyclopentyl 4-methoxyphenyl ketone

  • MDL number MFCD03841296
  • Cas Number : 15831-52-4
  • Molecular Weight : 204.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-03-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-03-07
Availability details for
Quantity
  • Availability
  • Add to Cart