RIE156323133 Rieke Metals, Inc.

4-(3-Oxocyclohexyl)butanenitrile

  • MDL number MFCD03841034
  • Cas Number : 110718-80-4
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6094C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6094C
Availability details for
Quantity
  • Availability
  • Add to Cart