RIE156323137 Rieke Metals, Inc.

3',2,2-Trimethylpropiophenone

  • MDL number MFCD03841163
  • Cas Number : 50390-49-3
  • Molecular Weight : 176.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6122-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6122-03
Availability details for
Quantity
  • Availability
  • Add to Cart