RIE156323146 Rieke Metals, Inc.

3'-Chloro-2-phenylacetophenone

  • MDL number MFCD02260665
  • Cas Number : 62482-45-5
  • Molecular Weight : 230.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6050-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6050-2
Availability details for
Quantity
  • Availability
  • Add to Cart