RIE156323149 Rieke Metals, Inc.

2'-bromo-2-phenylacetophenone

  • MDL number MFCD01861486
  • Cas Number : 36081-66-0
  • Molecular Weight : 275.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6051-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6051-1
Availability details for
Quantity
  • Availability
  • Add to Cart