RIE156323152 Rieke Metals, Inc.

2-chlorophenyl cyclopentyl ketone

  • MDL number MFCD00038367
  • Cas Number : 6740-85-8
  • Molecular Weight : 208.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6123-03-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-03-36
Availability details for
Quantity
  • Availability
  • Add to Cart