RIE156323155 Rieke Metals, Inc.

2,2-dimethyl-4'-thiomethylpropiophenone

  • MDL number MFCD00026313
  • Cas Number : 73242-19-0
  • Molecular Weight : 208.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6122-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6122-15
Availability details for
Quantity
  • Availability
  • Add to Cart