RIE156323157 Rieke Metals, Inc.

4'-Bromo-2,2-dimethylpropiophenone

  • MDL number MFCD03841175
  • Cas Number : 30314-45-5
  • Molecular Weight : 241.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6122-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6122-19
Availability details for
Quantity
  • Availability
  • Add to Cart