RIE156323167 Rieke Metals, Inc.

2,2,3',4'-tetramethylpropiophenone

  • MDL number MFCD03841183
  • Cas Number : 7397-00-4
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3,4-dimethylphenyl)-2,2-dimethyl-1-propanone
Inchi Code 1S/C13H18O/c1-9-6-7-11(8-10(9)2)12(14)13(3,4)5/h6-8H,1-5H3
Inchi Key DRXPDJIYWNCJPP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6122-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6122-30
Availability details for
Quantity
  • Availability
  • Add to Cart