RIE156323168 Rieke Metals, Inc.

4'-Methyl-2-phenylacetophenone

  • MDL number MFCD00017215
  • Cas Number : 2001-28-7
  • Molecular Weight : 210.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-methylphenyl)-2-phenylethanone
Inchi Code 1S/C15H14O/c1-12-7-9-14(10-8-12)15(16)11-13-5-3-2-4-6-13/h2-10H,11H2,1H3
Inchi Key RDBAEHHVVNBKBB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6053-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6053-3
Availability details for
Quantity
  • Availability
  • Add to Cart