RIE156323169 Rieke Metals, Inc.

2'-Methoxy-2-phenylacetophenone

  • MDL number MFCD02260671
  • Cas Number : 33470-10-9
  • Molecular Weight : 226.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6054-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6054-1
Availability details for
Quantity
  • Availability
  • Add to Cart