RIE156323171 Rieke Metals, Inc.

cyclohexyl 2-methylphenyl ketone

  • MDL number MFCD03841337
  • Cas Number : 2936-55-2
  • Molecular Weight : 202.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-04-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-02
Availability details for
Quantity
  • Availability
  • Add to Cart