RIE156323174 Rieke Metals, Inc.

cyclohexyl 2-methoxyphenyl ketone

  • MDL number MFCD03841339
  • Cas Number : 111504-19-9
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name cyclohexyl(2-methoxyphenyl)methanone
Inchi Code 1S/C14H18O2/c1-16-13-10-6-5-9-12(13)14(15)11-7-3-2-4-8-11/h5-6,9-11H,2-4,7-8H2,1H3
Inchi Key IOVWHDLNFVGIRP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6123-04-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-05
Availability details for
Quantity
  • Availability
  • Add to Cart