RIE156323177 Rieke Metals, Inc.

4'-Methoxy-2-phenylacetophenone

  • MDL number MFCD00017177
  • Cas Number : 1023-17-2
  • Molecular Weight : 226.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6054-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6054-3
Availability details for
Quantity
  • Availability
  • Add to Cart