RIE156323180 Rieke Metals, Inc.

4-chloro-1-(4-methylphenyl)-1-oxobutane

  • MDL number MFCD00018999
  • Cas Number : 38425-26-2
  • Molecular Weight : 196.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6104C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6104C
Availability details for
Quantity
  • Availability
  • Add to Cart