RIE156323183 Rieke Metals, Inc.

2-phenyl-3'-trifluoromethylacetophenone

  • MDL number MFCD02260677
  • Cas Number : 1533-04-6
  • Molecular Weight : 264.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6056-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6056-2
Availability details for
Quantity
  • Availability
  • Add to Cart