RIE156323186 Rieke Metals, Inc.

4-Bromophenyl cyclohexyl ketone

  • MDL number MFCD03841351
  • Cas Number : 3277-79-0
  • Molecular Weight : 267.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-04-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-19
Availability details for
Quantity
  • Availability
  • Add to Cart