RIE156323188 Rieke Metals, Inc.

4-Chlorophenyl cyclohexyl ketone

  • MDL number MFCD00454416
  • Cas Number : 3277-80-3
  • Molecular Weight : 222.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-04-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-21
Availability details for
Quantity
  • Availability
  • Add to Cart