RIE156323189 Rieke Metals, Inc.

4'-Nitro-2-phenylacetophenone

  • MDL number MFCD02260681
  • Cas Number : 3769-84-4
  • Molecular Weight : 241.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-nitrophenyl)-2-phenylethanone
Inchi Code 1S/C14H11NO3/c16-14(10-11-4-2-1-3-5-11)12-6-8-13(9-7-12)15(17)18/h1-9H,10H2
Inchi Key KWRIWTMLSQASGM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6057-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6057-3
Availability details for
Quantity
  • Availability
  • Add to Cart