RIE156323190 Rieke Metals, Inc.

Cyclohexyl 4-fluorophenyl ketone

  • MDL number MFCD03841354
  • Cas Number : 85014-02-4
  • Molecular Weight : 206.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-04-23

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-23
Availability details for
Quantity
  • Availability
  • Add to Cart