RIE156323193 Rieke Metals, Inc.

cyclohexyl 2,5-dimethylphenyl ketone

  • MDL number MFCD00447621
  • Cas Number : 2890-24-6
  • Molecular Weight : 216.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name cyclohexyl(2,5-dimethylphenyl)methanone
Inchi Code 1S/C15H20O/c1-11-8-9-12(2)14(10-11)15(16)13-6-4-3-5-7-13/h8-10,13H,3-7H2,1-2H3
Inchi Key DIXWFRFVEACSLS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6123-04-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-28
Availability details for
Quantity
  • Availability
  • Add to Cart