RIE156323194 Rieke Metals, Inc.

2-(3-Fluorophenyl)acetophenone

  • MDL number MFCD02260683
  • Cas Number : 347-90-0
  • Molecular Weight : 214.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3-fluorophenyl)-1-phenylethanone
Inchi Code 1S/C14H11FO/c15-13-8-4-5-11(9-13)10-14(16)12-6-2-1-3-7-12/h1-9H,10H2
Inchi Key UQGKOVXIHKXNPZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6058-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6058-2
Availability details for
Quantity
  • Availability
  • Add to Cart