RIE156323200 Rieke Metals, Inc.

2-(3-Chlorophenyl)acetophenone

  • MDL number MFCD02260685
  • Cas Number : 27798-43-2
  • Molecular Weight : 230.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6059-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6059-2
Availability details for
Quantity
  • Availability
  • Add to Cart