RIE156323201 Rieke Metals, Inc.

2-fluorophenyl cyclohexyl ketone

  • MDL number MFCD03841364
  • Cas Number : 106795-65-7
  • Molecular Weight : 206.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-04-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-37
Availability details for
Quantity
  • Availability
  • Add to Cart