RIE156323202 Rieke Metals, Inc.

2-(4-chlorophenyl)acetophenone

  • MDL number MFCD02260686
  • Cas Number : 6332-83-8
  • Molecular Weight : 230.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6059-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6059-3
Availability details for
Quantity
  • Availability
  • Add to Cart