RIE156323208 Rieke Metals, Inc.

2-(2-Methylphenyl)acetophenone

  • MDL number MFCD02260688
  • Cas Number : 5033-67-0
  • Molecular Weight : 210.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2-methylphenyl)-1-phenylethanone
Inchi Code 1S/C15H14O/c1-12-7-5-6-10-14(12)11-15(16)13-8-3-2-4-9-13/h2-10H,11H2,1H3
Inchi Key JMMXBSDTDHEQLI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6061-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6061-1
Availability details for
Quantity
  • Availability
  • Add to Cart