RIE156323211 Rieke Metals, Inc.

2-Valerylpyridine

  • MDL number MFCD07699185
  • Cas Number : 7137-97-5
  • Molecular Weight : 163.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5569-01C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-01C
Availability details for
Quantity
  • Availability
  • Add to Cart