RIE156323217 Rieke Metals, Inc.

2-(4-Methoxyphenyl)acetophenone

  • MDL number MFCD00096294
  • Cas Number : 24845-40-7
  • Molecular Weight : 226.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-methoxyphenyl)-1-phenylethanone
Inchi Code 1S/C15H14O2/c1-17-14-9-7-12(8-10-14)11-15(16)13-5-3-2-4-6-13/h2-10H,11H2,1H3
Inchi Key JABKESJVYSQBGF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6062-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6062-3
Availability details for
Quantity
  • Availability
  • Add to Cart