RIE156323223 Rieke Metals, Inc.

4-chlorophenyl cyclopropyl ketone

  • MDL number MFCD00001295
  • Cas Number : 6640-25-1
  • Molecular Weight : 180.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-01-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-01-21
Availability details for
Quantity
  • Availability
  • Add to Cart