RIE156323226 Rieke Metals, Inc.

6-Oxo-6-phenylhexanenitrile

  • MDL number MFCD02260532
  • Cas Number : 28353-03-9
  • Molecular Weight : 187.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6001D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6001D
Availability details for
Quantity
  • Availability
  • Add to Cart