RIE156323227 Rieke Metals, Inc.

cyclopropyl 2,4-dimethylphenyl ketone

  • MDL number MFCD00019224
  • Cas Number : 71172-79-7
  • Molecular Weight : 174.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name cyclopropyl(2,4-dimethylphenyl)methanone
Inchi Code 1S/C12H14O/c1-8-3-6-11(9(2)7-8)12(13)10-4-5-10/h3,6-7,10H,4-5H2,1-2H3
Inchi Key CENWGMGAFRZWTJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6123-01-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-01-27
Availability details for
Quantity
  • Availability
  • Add to Cart