RIE156323228 Rieke Metals, Inc.

cyclopropyl 2,5-dimethylphenyl ketone

  • MDL number MFCD00019225
  • Cas Number : 71172-44-6
  • Molecular Weight : 174.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-01-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-01-28
Availability details for
Quantity
  • Availability
  • Add to Cart