RIE156323231 Rieke Metals, Inc.

8-oxo-8-phenyloctanenitrile

  • MDL number MFCD02260534
  • Cas Number : 39755-15-2
  • Molecular Weight : 215.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6001F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6001F
Availability details for
Quantity
  • Availability
  • Add to Cart