RIE156323232 Rieke Metals, Inc.

4-(2-Bromophenyl)-4-oxobutyronitrile

  • MDL number MFCD02260535
  • Cas Number : 884504-61-4
  • Molecular Weight : 238.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6002B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6002B
Availability details for
Quantity
  • Availability
  • Add to Cart