RIE156323233 Rieke Metals, Inc.

5-(2-bromophenyl)-5-oxovaleronitrile

  • MDL number MFCD00672043
  • Cas Number : 884504-59-0
  • Molecular Weight : 252.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(2-bromophenyl)-5-oxopentanenitrile
Inchi Code 1S/C11H10BrNO/c12-10-6-2-1-5-9(10)11(14)7-3-4-8-13/h1-2,5-6H,3-4,7H2
Inchi Key MFTLRBDYXAYVIQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6002c

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6002c
Availability details for
Quantity
  • Availability
  • Add to Cart