RIE156323234 Rieke Metals, Inc.

6-(2-bromophenyl)-6-oxohexanenitrile

  • MDL number MFCD02260536
  • Cas Number : 884504-62-5
  • Molecular Weight : 266.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6002D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6002D
Availability details for
Quantity
  • Availability
  • Add to Cart