RIE156323238 Rieke Metals, Inc.

5-(3-bromophenyl)-5-oxovaleronitrile

  • MDL number MFCD00672044
  • Cas Number : 884504-60-3
  • Molecular Weight : 252.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6003C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6003C
Availability details for
Quantity
  • Availability
  • Add to Cart