RIE156323239 Rieke Metals, Inc.

Cyclopropyl 4-trifluoromethylphenyl ketone

  • MDL number MFCD03841231
  • Cas Number : 62587-07-9
  • Molecular Weight : 214.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name cyclopropyl[4-(trifluoromethyl)phenyl]methanone
Inchi Code 1S/C11H9F3O/c12-11(13,14)9-5-3-8(4-6-9)10(15)7-1-2-7/h3-7H,1-2H2
Inchi Key YNXJTEBLUUPSTE-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6123-01-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-01-45
Availability details for
Quantity
  • Availability
  • Add to Cart