RIE156323241 Rieke Metals, Inc.

Cyclohexylmethyl 2-pyridyl ketone

  • MDL number MFCD07699209
  • Cas Number : 149108-75-8
  • Molecular Weight : 203.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5569-08A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-08A
Availability details for
Quantity
  • Availability
  • Add to Cart