RIE156323245 Rieke Metals, Inc.

5-(2-iodophenyl)-5-oxovaleronitrile

  • MDL number MFCD02260587
  • Cas Number : 155589-54-1
  • Molecular Weight : 299.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6030C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6030C
Availability details for
Quantity
  • Availability
  • Add to Cart