RIE156323248 Rieke Metals, Inc.

Propyl picolinate

  • MDL number MFCD04338534
  • Cas Number : 98996-05-5
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5569-02B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-02B
Availability details for
Quantity
  • Availability
  • Add to Cart