RIE156323251 Rieke Metals, Inc.

4-chloro-1-(4-chlorophenyl)-1-oxobutane

  • MDL number MFCD00018993
  • Cas Number : 40877-09-6
  • Molecular Weight : 217.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6031C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6031C
Availability details for
Quantity
  • Availability
  • Add to Cart