RIE156323255 Rieke Metals, Inc.

6-Chloro-1-oxo-1-phenylhexane

  • MDL number MFCD00672047
  • Cas Number : 946-01-0
  • Molecular Weight : 210.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6005E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6005E
Availability details for
Quantity
  • Availability
  • Add to Cart