RIE156323258 Rieke Metals, Inc.

Isopropyl 2-pyridyl ketone

  • MDL number MFCD00778577
  • Cas Number : 59576-30-6
  • Molecular Weight : 149.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5569-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-10
Availability details for
Quantity
  • Availability
  • Add to Cart